Tantalum ≥99.9% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10348-G6 10348-H2
CAAA10348-G6EA 208.56 CAD
CAAA10348-G6 CAAA10348-H2
Tantalum ≥99.9% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)
Tantalum

MDL: MFCD00011252


Purity: 99.9+% CAS No.: 7440-25-7 Physical Form: ≈0.81g/m, Temper: soft, Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR