Tantalum (50 ppm Nb) ≥99.98% (metals basis), powder, Puratronic® -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

10344-14 10344-22
CAAA10344-14EA 243.32 CAD
CAAA10344-14 CAAA10344-22
Tantalum (50 ppm Nb) ≥99.98% (metals basis), powder, Puratronic® -100 mesh
Tantalum

MDL: MFCD00011252


Purity: 99.98% CAS No.: 7440-25-7 Appearance: Black Odor: Odorless UN No.: 3089 MDL No.: MFCD00011252

Formula: Ta
Density: 16,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR