Sodium ≥99.8% (metals basis), ingot

Supplier: Thermo Scientific Chemicals
Danger

10342-35 10342-60
CAAA10342-35EA 206.98 CAD
CAAA10342-35 CAAA10342-60
Sodium ≥99.8% (metals basis), ingot
Sodium

MDL: MFCD00085307 Notes: Product meets ACS specifications


Purity: 99.8% CAS No.: 7440-23-5 Appearance: Silver-white Odor: Odorless UN No.: 1428 MDL No.: MFCD00085307

Formula: Na
Boiling Pt: 881 °C (1013 hPa)
Melting Pt: 97.8 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085307
CAS Number: 7440-23-5
EINECS: 231-132-9
UN: 1428
ADR: 4.3,I
Merck Index: 13,08641

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR