Sodium ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

10336-03 10336-06
CAAA10336-03EA 189.6 CAD
CAAA10336-03 CAAA10336-06
Sodium ≥99.95% (metals basis)
Sodium

MDL: MFCD00085307


Purity: 99.95% CAS No.: 7440-23-5 Physical Form: Sealed in breakseal ampoules Odor: Odorless UN No.: 1428 MDL No.: MFCD00085307

Formula: Na
Boiling Pt: 881 °C (1013 hPa)
Melting Pt: 97.8 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085307
CAS Number: 7440-23-5
EINECS: 231-132-9
UN: 1428
ADR: 4.3,I
Merck Index: 13,08641

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR