Silicon ≥98.4% (metals basis), lumps 10 cm (3.9 in) & down

Supplier: Thermo Scientific Chemicals
Warning

10335-A1 10335-A7
CAAA10335-A1EA 103.33 CAD
CAAA10335-A1 CAAA10335-A7
Silicon ≥98.4% (metals basis), lumps 10 cm (3.9 in) & down
Silicon

MDL: MFCD00085311


Purity: 98.4% CAS No.: 7440-21-3 Odor: Odorless MDL No.: MFCD00085311

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR