Rhenium ≥99.99% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
10313-04
10313-09
10313-18
10313-SS
CAAA10313-04EA
229.1
CAD
CAAA10313-04
CAAA10313-09
CAAA10313-18
CAAA10313-SS
Rhenium ≥99.99% (metals basis), powder -325 mesh
Rhenium
MDL: MFCD00011195
Purity: 99.99%
CAS No.: 7440-15-5
Appearance: Black to grey
UN No.: 3089
MDL No.: MFCD00011195
Formula:
Re MW: 186.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...