Rhenium ≥99.97% (metals basis), wire, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

10312-BQ 10312-BU 10312-BY
CAAA10312-BQEA 109.81 CAD
CAAA10312-BQ CAAA10312-BU CAAA10312-BY
Rhenium ≥99.97% (metals basis), wire, Ø 0.5 mm (0.02 in)
Rhenium

MDL: MFCD00011195


Purity: 99.97% CAS No.: 7440-15-5 Physical Form: ≈4.13g/m Odor: Odorless MDL No.: MFCD00011195

Formula: Re
Boiling Pt: 5630 °C (1013 hPa)
Melting Pt: 3180 °C
Density: 21.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
Merck Index: 12,08343

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR