Diammonium hydrogen orthophosphate P₂O₅ ≥ 53%, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
10301-A1
10301-A7
CAAA10301-A1EA
122.29
CAD
CAAA10301-A1
CAAA10301-A7
Diammonium hydrogen orthophosphate P₂O₅ ≥ 53%, tech.
di-Ammonium hydrogen orthophosphate
Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc
CAS No.: 7783-28-0
Molecular Formula: (NH4)2HPO4
Formula Weight: 132.06
Physical Form: -100 Mesh Powder
Density: 1.619
Appearance: White
Melting Point: 155° dec.
Solubility: Freely soluble in water. Insoluble in alcohol, acetone
Merck Reference: 14,542
MDL No.: MFCD00010891
Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc
Formula:
(NH₄)₂HPO₄ Melting Pt: 155 °C (dec.) Density: 1.619 Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 EINECS: 231-987-8 REACH: 01-2119490974-22 Merck Index: 14,542 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...