Potassium ≥99.95% (metals basis), solid

Supplier: Thermo Scientific Chemicals
Danger

10299-03 10299-06
CAAA10299-03EA 255.96 CAD
CAAA10299-03 CAAA10299-06
Potassium ≥99.95% (metals basis), solid
Potassium

MDL: MFCD00133776


Purity: 99.95% CAS No.: 7440-09-7 Physical Form: Sealed in standard ampoules under vacuum Odor: Odorless UN No.: 2257 MDL No.: MFCD00133776

Formula: K
MW: 39.1
Boiling Pt: 760
Melting Pt: 63
Density: 0.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133776
CAS Number: 7440-09-7
EINECS: 231-119-8
UN: 2257
ADR: 4.3 I
Merck Index: 12,07763

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR