Platinum, wire for ISA type R or S standard grade thermocouple, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
10288-BU
10288-BY
10288-CC
CAAA10288-BUEA
186.44
CAD
CAAA10288-BU
CAAA10288-BY
CAAA10288-CC
Platinum, wire for ISA type R or S standard grade thermocouple, Ø 0.25 mm (0.01 in)
Platinum
For ISA Type R or S Standard Grade Thermocouple
CAS No.: 7440-06-4
Formula Weight: 195.08
Physical Form: ≈1.09g/m
Density: 21.45G/CC
Odor: Odorless
Melting Point: 1768.4 deg C
Boiling Point: 3825 deg C
MDL No.: MFCD00011179
Formula:
Pt Boiling Pt: 3825 °C Melting Pt: 1768.4 °C Density: 21.45G/CC Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...