Platinum ≥99.95% (metals basis), wire, hard, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

10286-BU 10286-BY
CAAA10286-BUEA 657.28 CAD
CAAA10286-BU CAAA10286-BY
Platinum ≥99.95% (metals basis), wire, hard, Ø 0.5 mm (0.02 in)
Platinum

MDL: MFCD00011179


Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈4.21g/m, Temper: as drawn Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR