Platinum 99,95 Metals basis, wire, hard, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Platinium
10285-BQ
10285-BU
CAAA10285-BQEA
475.58
CAD
CAAA10285-BQ
CAAA10285-BU
Platinum 99,95 Metals basis, wire, hard, Ø 1.0 mm (0.04 in)
Platinum
MDL: MFCD00011179
Purity: 99.95%
CAS No.: 7440-06-4
Physical Form: ≈16.8g/m, Temper: as drawn
Odor: Odorless
MDL No.: MFCD00011179
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Density: 21.447 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...