Palladium ≥99.9%, wire, Ø 1.0 mm

Supplier: Thermo Scientific Chemicals

10280-BS 10280-BW
CAAA10280-BSEA 492.96 CAD
CAAA10280-BS CAAA10280-BW
Palladium ≥99.9%, wire, Ø 1.0 mm
Palladium

MDL: MFCD00011167


Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈9.44g/m Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR