Palladium ≥99.9%, wire, Ø 0.5 mm

Supplier: Thermo Scientific Chemicals

10279-BW 10279-CB 10279-CD
CAAA10279-BWEA 616.2 CAD
CAAA10279-BW CAAA10279-CB CAAA10279-CD
Palladium ≥99.9%, wire, Ø 0.5 mm
Palladium

MDL: MFCD00011167


Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈2.36g/m Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR