Niobium ≥99.8% (metals basis excluding Ta), powder -60 mesh (max. 1% Ta)
Supplier: Thermo Scientific Chemicals
10276-14
10276-22
10276-36
CAAA10276-14EA
66.36
CAD
CAAA10276-14
CAAA10276-22
CAAA10276-36
Niobium ≥99.8% (metals basis excluding Ta), powder -60 mesh (max. 1% Ta)
Niobium
MDL: MFCD00011126
Purity: 99.8%
CAS No.: 7440-03-1
Odor: Odorless
MDL No.: MFCD00011126
Formula:
Nb Storage Temperature: Ambient |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...