Niobium ≥99.8% (metals basis), wire, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
10264-G1
10264-G5
10264-G9
CAAA10264-G1EA
76.16
CAD
CAAA10264-G1
CAAA10264-G5
CAAA10264-G9
Niobium ≥99.8% (metals basis), wire, Ø 1.0 mm (0.04 in)
Niobium
MDL: MFCD00011126
Purity: 99.8%
CAS No.: 7440-03-1
Physical Form: ≈6.7g/m
Odor: Odorless
MDL No.: MFCD00011126
Formula:
Nb Boiling Pt: 3700 °C (1013 hPa) Melting Pt: 2468 °C Density: 8.57 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...