Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10259-FY 10259-GK 10259-HQ
CAAA10259-FYEA 76.47 CAD
CAAA10259-FY CAAA10259-GK CAAA10259-HQ
Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.25 mm (0.01 in)
Niobium

MDL: MFCD00011126


Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈10.71g/50x100mm Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR