Nickel ≥99.9% (metals basis), powder APS 3-7 µm

Supplier: Thermo Scientific Chemicals
Danger

10256-22 10256-36 10256-A3
CAAA10256-22EA 78.53 CAD
CAAA10256-22 CAAA10256-36 CAAA10256-A3
Nickel ≥99.9% (metals basis), powder APS 3-7 µm
Nickel

MDL: MFCD00011137


Purity: 99.9% CAS No.: 7440-02-0 Physical Form: Apparent density 1.8-2.7g/cm3 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011137

Formula: Ni
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR