Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

10248-FF 10248-FI
CAAA10248-FFEA 358.66 CAD
CAAA10248-FF CAAA10248-FI
Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Neodymium

MDL: MFCD00011130


Purity: 99.9% CAS No.: 7440-00-8 Physical Form: ≈4.4g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011130

Formula: Nd
MW: 144.24
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR