Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.3 mm (0.012 in)

Supplier: Thermo Scientific Chemicals

10247-FF 10247-FI
CAAA10247-FFEA 385.52 CAD
CAAA10247-FF CAAA10247-FI
Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.3 mm (0.012 in)
Neodymium

MDL: MFCD00011130


Purity: 99.9% CAS No.: 7440-00-8 Physical Form: ≈1.3g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011130

Formula: Nd
MW: 144.24
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR