Manganese ≥99.3% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

10238-14 10238-30 10238-A1
CAAA10238-14EA 44.08 CAD
CAAA10238-14 CAAA10238-30 CAAA10238-A1
Manganese ≥99.3% (metals basis), powder -325 mesh
Manganese

MDL: MFCD00011111


Purity: 99.3% CAS No.: 7439-96-5 Physical Form: Apparent density 2.6-3.5g/cm3 UN No.: 3089 MDL No.: MFCD00011111

Formula: Mn
MW: 54.94 g/mol
Boiling Pt: 2097 °C (1013 hPa)
Melting Pt: 1260 °C
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,05762

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR