Magnesium ≥99.8% (metals basis), turnings

Supplier: Thermo Scientific Chemicals
Danger

10235-22 10235-36 10235-57
CAAA10235-22EA 86.9 CAD
CAAA10235-22 CAAA10235-36 CAAA10235-57
Magnesium ≥99.8% (metals basis), turnings
Magnesium

MDL: MFCD00085308


Purity: 99.8% Physical Form: 2.3mm (0.09in) wide Appearance: Silver-grey Odor: Odorless UN No.: 1869 MDL No.: MFCD00085308

Formula: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR