Magnesium ≥99.8% (metals basis), rods, Length: 30 cm (12 in), Ø 3.3 cm (1.3 in)

Supplier: Thermo Scientific Chemicals

10231-KG 10231-KJ 10231-KM
CAAA10231-KGEA 98.75 CAD
CAAA10231-KG CAAA10231-KJ CAAA10231-KM
Magnesium ≥99.8% (metals basis), rods, Length: 30 cm (12 in), Ø 3.3 cm (1.3 in)
Magnesium

MDL: MFCD00085308


Purity: 99.8% CAS No.: 7439-95-4 Physical Form: ≈450g/pc Odor: Odorless MDL No.: MFCD00085308

Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR