Lead : Antimony (94:6)%, shot 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Danger

10224-36 10224-A3 10224-A9
CAAA10224-36EA 77.89 CAD
CAAA10224-36 CAAA10224-A3 CAAA10224-A9
Lead : Antimony (94:6)%, shot 2 mm (0.08 in)
Lead : Antimony

MDL: MFCD00134050


Odor: Odorless UN No.: 3077 MDL No.: MFCD00134050

Formula: Sb
Boiling Pt: 1637 °C (1013 hPa)
Melting Pt: 630 °C
Density: 6.684 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7440-36-0
EINECS: 231-100-4
UN: 3077
ADR: 9,III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR