Iron ≥99.97% (metals basis), pieces, irregularly shaped ≤ 25 mm (1.0 in)

Supplier: Thermo Scientific Chemicals

10213-30 10213-A1 10213-A7
CAAA10213-30EA 73.47 CAD
CAAA10213-30 CAAA10213-A1 CAAA10213-A7
Iron ≥99.97% (metals basis), pieces, irregularly shaped ≤ 25 mm (1.0 in)
Iron

MDL: MFCD00010999


Purity: 99.97+% CAS No.: 7439-89-6 Physical Form: 90% 2.5-25mm (0.1-1.0in) Appearance: Gray MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR