Holmium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.3 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10210-FF 10210-FI
CAAA10210-FFEA 453.46 CAD
CAAA10210-FF CAAA10210-FI
Holmium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.3 mm (0.01 in)
Holmium

MDL: MFCD00011049


Purity: 99.9% CAS No.: 7440-60-0 Physical Form: ≈1.65g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011049

Formula: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR