Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

10205-BU 10205-BY 10205-CC
CAAA10205-BUEA 96.38 CAD
CAAA10205-BU CAAA10205-BY CAAA10205-CC
Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 1.0 mm (0.04 in)
Hafnium

MDL: MFCD00011032


Purity: 99.95% CAS No.: 7440-58-6 Physical Form: ≈10.28g/m Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR