Hafnium ≥99.6% (trace metals basis excluding Zr), powder -325 mesh (max. 2-3.5% Zr)

Supplier: Thermo Scientific Chemicals
Danger

10201-06 10201-14 10201-22
CAAA10201-06EA 118.5 CAD
CAAA10201-06 CAAA10201-14 CAAA10201-22
Hafnium ≥99.6% (trace metals basis excluding Zr), powder -325 mesh (max. 2-3.5% Zr)
Hafnium

MDL: MFCD00011032


Purity: 99.6% CAS No.: 7440-58-6 Appearance: Grey Odor: Odorless UN No.: 2545 MDL No.: MFCD00011032

Formula: Hf
MW: 178.49 g/mol
Boiling Pt: 5400 °C (1013 hPa)
Melting Pt: 2224 °C
Density: 13.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4
UN: 2545
ADR: 4.2,II
Merck Index: 12,04618

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR