Hafnium ≥99.97% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10200-G2 10200-G6
CAAA10200-BWEA 228.75 CAD
CAAA10200-BW CAAA10200-G2 CAAA10200-G6
Hafnium ≥99.97% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.25 mm (0.01 in)
Hafnium

MDL: MFCD00011032


Purity: 99.97% CAS No.: 7440-58-6 Physical Form: ≈0.64g/m Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR