Germanium ≥99.999% (metals basis), pieces 3-9 mm (0.1-0.4 in)
Supplier: Thermo Scientific Chemicals
10191-09
10191-18
CAAA10191-09EA
346.02
CAD
CAAA10191-09
CAAA10191-18
Germanium ≥99.999% (metals basis), pieces 3-9 mm (0.1-0.4 in)
Germanium
MDL: MFCD00085310
Purity: 99.999%
CAS No.: 7440-56-4
Odor: Odorless
MDL No.: MFCD00085310
Formula:
Ge Boiling Pt: 2830 °C (1013 hPa) Melting Pt: 937.2 °C Density: 5.323 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 Merck Index: 12,04415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...