Gallium ≥99.999% (metals basis), ingot

Supplier: Thermo Scientific Chemicals
Warning

10187-30 10187-36
CAAA10187-30EA 2066.64 CAD
CAAA10187-30 CAAA10187-36
Gallium ≥99.999% (metals basis), ingot
Gallium

MDL: MFCD00134045


Purity: 99.999% CAS No.: 7440-55-3 Molecular Formula: Requires overnight shipping with cold pack Storage Temperature: 2°C to 8°C Appearance: Silver grey MDL No.: MFCD00134045

Formula: Ga
MW: 69.72 g/mol
Boiling Pt: 2403
Melting Pt: 29
Density: 5.904 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8 III
Merck Index: 12,04364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR