Tri-tert-butylphosphine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10178-03
10178-06
CAAA10178-03EA
219.62
CAD
CAAA10178-03
CAAA10178-06
Tri-tert-butylphosphine 98%
Tri-tert-butylphosphine
MDL: MFCD00015006
Purity: 96%
CAS No.: 13716-12-6
Molecular Formula: C12H27P
Formula Weight: 202.32
Flash Point: -16°(-3°F)
Physical Form: Solid
Sensitivity: Air and Moisture Sensitive
Appearance: White
Odor: Pungent
Melting Point: 30°
Boiling Point: 102-103°/13mm
UN No.: 2846
MDL No.: MFCD00015006
Formula:
C₁₂H₂₇P Boiling Pt: 102…103 °C (13 mmHg) Melting Pt: 30 °C Flash Pt: –16 °C (3 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015006 CAS Number: 13716-12-6 EINECS: 237-266-4 UN: 2846 ADR: 4.2,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...