Copper 99+% (metals basis), turnings

Supplier: Thermo Scientific Chemicals

10161-18 10161-30 10161-A1
CAAA10161-18EA 33.65 CAD
CAAA10161-18 CAAA10161-30 CAAA10161-A1
Copper 99+% (metals basis), turnings
Copper

MDL: MFCD00010965


Purity: 99+% CAS No.: 7440-50-8 Physical Form: Long, thin, coiled strands Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR