Copper ≥99.5% (metals basis), powder -150 mesh

Supplier: Thermo Scientific Chemicals
Warning

10160-22 10160-36 10160-A3
CAAA10160-22EA 73 CAD
CAAA10160-22 CAAA10160-36 CAAA10160-A3
Copper ≥99.5% (metals basis), powder -150 mesh
Copper

MDL: MFCD00010965


Purity: 99.5% CAS No.: 7440-50-8 Molecular Formula: Apparent density 2.6-2.8g/cm3 Appearance: Copper Odor: Odorless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR