Chromium ≥99.997% (metals basis), pieces, Puratronic® ≤ 6 mm electrolytic solution

Supplier: Thermo Scientific Chemicals

10151-14 10151-22
CAAA10151-14EA 152.15 CAD
CAAA10151-14 CAAA10151-22
Chromium ≥99.997% (metals basis), pieces, Puratronic® ≤ 6 mm electrolytic solution
Chromium

MDL: MFCD00010944


Purity: 99.997% CAS No.: 7440-47-3 Odor: Odorless MDL No.: MFCD00010944

Formula: Cr
Boiling Pt: 2469 °C (1013 hPa)
Melting Pt: 1840 °C
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR