Chromium ≥99% (metals basis), irregular pieces

Supplier: Thermo Scientific Chemicals

10150-22 10150-36
CAAA10150-22EA 113.44 CAD
CAAA10150-22 CAAA10150-36
Chromium ≥99% (metals basis), irregular pieces
Chromium

MDL: MFCD00010944


Purity: 99% CAS No.: 7440-47-3 Physical Form: Broken plate Odor: Odorless MDL No.: MFCD00010944

Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR