Chromium ≥99.97% (metals basis), cubes, Length: 12.5 mm (0.492 in)

Supplier: Thermo Scientific Chemicals

10149-KT 10149-KU
CAAA10149-KTEA 289.14 CAD
CAAA10149-KT CAAA10149-KU
Chromium ≥99.97% (metals basis), cubes, Length: 12.5 mm (0.492 in)
Chromium

MDL: MFCD00010944


Purity: 99.97% CAS No.: 7440-47-3 Physical Form: ≈14.7g/pc Hot Isostatically Pressed Odor: Odorless MDL No.: MFCD00010944

Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR