Chromium ≥99% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

10148-22 10148-36 10148-A3
CAAA10148-22EA 91.64 CAD
CAAA10148-22 CAAA10148-36 CAAA10148-A3
Chromium ≥99% (metals basis), powder -325 mesh
Chromium

MDL: MFCD00010944


Purity: 99% CAS No.: 7440-47-3 Odor: Odorless UN No.: 3089 MDL No.: MFCD00010944

Formula: Cr
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR