Cadmium ≥99.95% (metals basis), tear drop shot -5/+20 mesh

Supplier: Thermo Scientific Chemicals
Danger

10122-18 10122-30 10122-A1
CAAA10122-18EA 71.1 CAD
CAAA10122-18 CAAA10122-30 CAAA10122-A1
Cadmium ≥99.95% (metals basis), tear drop shot -5/+20 mesh
Cadmium

MDL: MFCD00010914


Purity: 99.95% CAS No.: 7440-43-9 MDL No.: MFCD00010914

Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR