Cadmium ≥99.999% (metals basis), rods, Puratronic® 23 cm (9.0 in), Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals
Danger

10120-KG 10120-KJ
CAAA10120-KGEA 478.74 CAD
CAAA10120-KG CAAA10120-KJ
Cadmium ≥99.999% (metals basis), rods, Puratronic® 23 cm (9.0 in), Ø 12.7 mm (0.5 in)
Cadmium

MDL: MFCD00010914


Purity: 99.999+% CAS No.: 7440-43-9 Physical Form: ≈250g/rod MDL No.: MFCD00010914

Formula: Cd
MW: 112.41 g/mol
Boiling Pt: 765
Melting Pt: 321
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1 II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR