Cadmium ≥99.99% (metals basis), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals
Danger

10119-UH 10119-UI 10119-UT
CAAA10119-UHEA 357.08 CAD
CAAA10119-UH CAAA10119-UI CAAA10119-UT
Cadmium ≥99.99% (metals basis), rods, Ø 12.7 mm (0.5 in)
Cadmium

MDL: MFCD00010914


Purity: 99.99+% CAS No.: 7440-43-9 Physical Form: ≈11.0g/cm MDL No.: MFCD00010914

Formula: Cd
MW: 112.41 g/mol
Boiling Pt: 765
Melting Pt: 321
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1 II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR