Bismuth ≥99.5% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

10111-22 10111-36
CAAA10111-22EA 101.12 CAD
CAAA10111-22 CAAA10111-36
Bismuth ≥99.5% (metals basis), powder -325 mesh
Bismuth

MDL: MFCD00134033


Purity: 99.5% CAS No.: 7440-69-9 Physical Form: Apparent density 3.2-4.0g/cm3 Odor: Odorless UN No.: 3089 MDL No.: MFCD00134033

Formula: Bi
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR