Bismuth 99,5 Metals basis, powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

10110-22 10110-36 10110-A3
CAAA10110-22EA 84.69 CAD
CAAA10110-22 CAAA10110-36 CAAA10110-A3
Bismuth 99,5 Metals basis, powder -100 mesh
Bismuth

MDL: MFCD00134033


Purity: 99.5% CAS No.: 7440-69-9 Physical Form: Apparent density 3.8-4.6g/cm3 Odor: Odorless UN No.: 3089 MDL No.: MFCD00134033

Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR