Barium 99+% (metals basis), rods (max. 0.8% Sr), Ø 22 mm (0.9 in)

Supplier: Thermo Scientific Chemicals
Danger


Packaged under argon.

10103-22 10103-36 10103-A1
CAAA10103-22EA 319.16 CAD
CAAA10103-22 CAAA10103-36 CAAA10103-A1
Barium 99+% (metals basis), rods (max. 0.8% Sr), Ø 22 mm (0.9 in)
Barium

Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps


Purity: 99+% CAS No.: 7440-39-3 Physical Form: ≈7.3cm/100g, Packaged under argon Appearance: Grey Odor: Odorless UN No.: 1400 MDL No.: MFCD00134031


Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps

Formula: Ba
Boiling Pt: 1640 °C (1013 hPa)
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR