Arsenic ≥99% (metals basis), powder -20 mesh

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Arsenic solution

10101-18 10101-30
CAAA10101-18EA 243.32 CAD
CAAA10101-18 CAAA10101-30
Arsenic ≥99% (metals basis), powder -20 mesh
Arsenic

MDL: MFCD00085309


Purity: 99% CAS No.: 7440-38-2 Appearance: Silver to black Odor: Odorless UN No.: 1558 MDL No.: MFCD00085309

Formula: As
MW: 74.92 g/mol
Density: 5,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085309
CAS Number: 7440-38-2
EINECS: 213-148-6
UN: 1558
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR