Antimony ≥99.5% (metals basis), powder -100 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
10100-30
10100-A1
CAAA10100-30EA
76.79
CAD
CAAA10100-30
CAAA10100-A1
Antimony ≥99.5% (metals basis), powder -100 mesh
Antimony
MDL: MFCD00134030
Purity: 99.5%
CAS No.: 7440-36-0
Physical Form: Apparent density 2.4-2.9g/cm3
Odor: Odorless
Boiling Point: 1380°
UN No.: 2871
MDL No.: MFCD00134030
Formula:
Sb Boiling Pt: 1380 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...