Antimony ≥99.5% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

10099-09 10099-22 10099-36 10099-A3
CAAA10099-09EA 38.39 CAD
CAAA10099-09 CAAA10099-22 CAAA10099-36 CAAA10099-A3
Antimony ≥99.5% (metals basis), powder -200 mesh
Antimony

MDL: MFCD00134030


Purity: 99.5% CAS No.: 7440-36-0 Appearance: Gray Odor: Odorless UN No.: 2871 MDL No.: MFCD00134030

Formula: Sb
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR