Antimony ≥99.5% (metals basis), lumps ≤ 1.25 cm (0.49 in)

Supplier: Thermo Scientific Chemicals
Warning

10095-22 10095-36 10095-A3
CAAA10095-22EA 56.56 CAD
CAAA10095-22 CAAA10095-36 CAAA10095-A3
Antimony ≥99.5% (metals basis), lumps ≤ 1.25 cm (0.49 in)
Antimony

MDL: MFCD00134030


Purity: 99.5% CAS No.: 7440-36-0 Odor: Odorless MDL No.: MFCD00134030

Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637 °C (1013 hPa)
Melting Pt: 630 °C
Density: 6.684 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR