Platinum iridium alloy (90:10 w%), wire, hard, Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
10056-BS
10056-BW
10056-CB
CAAA10056-BSEA
227.52
CAD
CAAA10056-BS
CAAA10056-BW
CAAA10056-CB
Platinum iridium alloy (90:10 w%), wire, hard, Ø 0.5 mm (0.02 in)
Platinum Iridium alloy (90:10 w%)
MDL: MFCD02091683
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.44g/10cm, Temper: as drawn
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...