Platinum iridium alloy (80:20 w%), wire, hard, Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
10054-BS
10054-BW
CAAA10054-BSEA
211.72
CAD
CAAA10054-BS
CAAA10054-BW
Platinum iridium alloy (80:20 w%), wire, hard, Ø 0.5 mm (0.02 in)
Platinum Iridium alloy (80:20 w%)
MDL: MFCD02091683
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.44g/10cm, Temper: as drawn
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...